missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rae-1 Antibody (199205), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB17581Y
This item is not returnable.
View return policy
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry
Specifications
| Rae-1 | |
| Monoclonal | |
| DyLight 680 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8480 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 199205 | |
| 50mM Sodium Borate | |
| BaF3 mouse pro-B cell line transfected with Rae-1 beta (Accession # NP_033043) and Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta Leu29-Ser227 (Accession # Q9JI58) | |
| 0.5 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction