Learn More
Rae-1 epsilon Antibody (205001) [DyLight 350], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1135D
This item is not returnable.
View return policy
Description
Rae-1 epsilon Monoclonal antibody specifically detects Rae-1 epsilon in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Specifications
| Rae-1 epsilon | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| BaF3 mouse pro-B cell line transfected with mouse Rae-1 epsilon | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 205001 | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Mouse | |
| Purified |