missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rae-1 Rat anti-Mouse, BUV395, Clone: 186107, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 748218
This item is not returnable.
View return policy
Description
Rae-1 Monoclonal antibody specifically detects Rae-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Rae-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Rae-1 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 186107 | |
| Brilliant Ultraviolet 395 | |
| 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction