missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAE1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1894
This item is not returnable.
View return policy
Description
RAE1 Polyclonal specifically detects RAE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| RAE1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| dJ481F12.3, dJ800J21.1, FLJ30608, homolog of yeast Rae1 (Bharathi) mRNA-associated protein of 41 kDa (Kraemer), MGC117333, MGC126076, MGC126077, MIG14, migration-inducing gene 14, Mnrp41, mRNA export factor, mRNA export protein, mRNA-associated protein mrnp 41, mRNA-binding protein, 41-kD, MRNP41, RAE1 (RNA export 1, S.pombe) homolog, Rae1 protein homolog, RAE1 RNA export 1 homolog (S. pombe) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8480 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 3 - 6 ug/ml, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_003601.1 | |
| RAE1 | |
| Peptide with sequence C-NAAEELKPRNKK corresponding to C-Terminus according to NP_003601.1, NP_001015885.1. | |
| 0.1 mg | |
| Primary | |
| NP_003601and NP_001015885 represent identical proteins. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction