missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raf-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23356
This item is not returnable.
View return policy
Description
Raf-1 Polyclonal antibody specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Raf-1 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| cRaf, c-Raf, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf-1Oncogene RAF1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
| Raf-1 synthetic peptide (coupled to KLH) corresponds to amino acids from the C-terminus of human Raf-1. This sequence is conserved in Raf-1 from many species, and is not found in A-Raf or B-Raf. [UniProt# P04049] | |
| 0.1 mL | |
| Apoptosis, Cancer, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
| 5894 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction