Learn More
Invitrogen™ RAG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585711
Description
Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (95%), Xenopus laevis (84%), Pig (90%), Rabbit (96%), Chicken (86%), Rhesus Monkey (98%). Positive Control: Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is involved in activation of immunoglobulin V-D-J recombination. The encoded protein is involved in recognition of the DNA substrate, but stable binding and cleavage activity also requires RAG2. Defects in this gene can be the cause of several diseases.
Specifications
| RAG1 | |
| Polyclonal | |
| Unconjugated | |
| Rag1 | |
| E3 ubiquitin-protein ligase RAG1; Endonuclease RAG1; MGC43321; Rag1; RAG-1; RAG-1 Antibody; recombinase activating gene 1; recombinase activating protein; recombination activating 1; recombination activating gene 1; recombination activating gene 1 Antibody; recombination activating gene-1; recombination activating protein 1; recombination activation protein 1; recombination-activating gene 1; RING finger protein 74; RING-type E3 ubiquitin transferase RAG1; RNF74; RNF74 Antibody; V(D)J recombination-activating protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5896 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.85 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P15918 | |
| Rag1 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human RAG1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.