missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAGE Mouse anti-Human, Mouse, Clone: OTI6C12, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810865S
This item is not returnable.
View return policy
Description
RAGE Monoclonal antibody specifically detects RAGE in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RAGE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15109, Q62151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 11596, 177 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C12 | |
| Unconjugated | |
| AGER | |
| advanced glycation end product receptor; advanced glycation end-product receptor; advanced glycation end-products receptor; advanced glycosylation end product-specific receptor; advanced glycosylation end product-specific receptor variant 2; advanced glycosylation end product-specific receptor variant 3; advanced glycosylation end product-specific receptor variant 4; advanced glycosylation end product-specific receptor variant 5; advanced glycosylation end-product specific receptor; Ager; MAPK/MAK/MRK overlapping kinase; MOK; MOK protein kinase; RAGE; RAGE isoform NtRAGE-delta; RAGE isoform sRAGE-delta; RAGE/AGER; RAGE1; RAGE-1; RAGE-4 ORF3; receptor for advanced glycation endproducts; receptor for advanced glycation end-products variant 20; receptor for advanced glycosylation end products; receptor of advanced glycosylation end products of proteins; immunoglobulin superfamily; MHC class II; MHC class III; renal cell carcinoma antigen; renal tumor antigen 1; SCARJ1; sRAGE; STK30 | |
| Human recombinant protein fragment corresponding to amino acids 205-419 of human RAGE produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction