missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAI16 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97743PE
This item is not returnable.
View return policy
Description
RAI16 Polyclonal antibody specifically detects RAI16 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| RAI16 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| family with sequence similarity 160, member B2, FLJ11125, FLJ21801, MGC138352, RAI16, retinoic acid induced 16, Retinoic acid-induced protein 16 | |
| Produced in rabbits immunized with E. coli-derived Human RAI16 fragment. | |
| 0.1 mL | |
| Cell Biology | |
| 64760 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction