missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAI3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110739
This item is not returnable.
View return policy
Description
Immunogen sequence: LTKQRNPMDY PVEDAFCKPQ LVKKSYGVEN RAYSQEEITQ GFEETGDTLY APYSTHFQLQ NQPPQKEFSI PR Highest antigen sequence indentity to the following orthologs: Rat - 81%, Mouse - 82%.
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function for this retinoic acid-induced gene has not yet been determined; however, it may play a role in embryonic development and epithelial cell differentiation. This gene is a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. This gene may also be involved in interaction between retinoid acid and G protein signalling pathways. RAI3 has been reported in lung, kidney, colon, pancreas, prostate, and small intestine. ESTs have been isolated from amnion, breast, colon, eye, lung, nasal cavity, placenta, skin, uterus, and vessel libraries.
Specifications
| RAI3 | |
| Polyclonal | |
| Unconjugated | |
| GPRC5A | |
| G protein-coupled receptor class C group 5 member A; G protein-coupled receptor, class C, group 5, member A; G protein-coupled receptor, family C, group 5, member A; GPCR5A; Gprc5a; G-protein coupled receptor family C group 5 member A; orphan G-protein-coupling receptor PEIG-1; PEIG-1; Phorbol ester induced gene 1; phorbol ester induced protein-1; Rai3; Raig1; RAIG-1; retinoic acid induced 3; retinoic acid responsive; retinoic acid-induced gene 1 protein; Retinoic acid-induced protein 3; TIG1; TPA induced gene 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9052 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFJ5 | |
| GPRC5A | |
| Recombinant Protein corresonding to Human RAI3. Recombinant protein control fragment (Product #RP-89541). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction