missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAIDD/CRADD Antibody (14G8), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42688AF647
This item is not returnable.
View return policy
Description
RAIDD/CRADD Monoclonal specifically detects RAIDD/CRADD in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| RAIDD/CRADD | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CRADD | |
| Recombinant human RAIDD/CRADD (1-199aa) purified from E. coli | |
| 100 μL | |
| Apoptosis, Cancer, Cell Biology, Tumor Suppressors | |
| 8738 | |
| Store at 4C in the dark. | |
| IgG1a κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 14G8 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| CASP2 and RIPK1 domain containing adaptor with death domain, Caspase and RIP adapter with death domain, caspase and RIP adaptor with death domain, death domain containing protein CRADD, death domain-containing protein CRADD, MGC9163, RAIDDdeath adaptor molecule RAIDD, RIP-associated ICH1/CED3-homologous protein with death domain, RIP-associated protein with a death domain | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction