Learn More
Mettler Toledo™ Rainin™ BioClean™ Bulk Pipette Tips
Product Code. 12029011
Encompass
Change view
Click to view available options
Volume (Metric):
300 μL
1000 μL
2000 μL
Unit Size:
Pack of 1000
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12029011 300 μL Pack of 1000
12120581 2000 μL Pack of 1000
12099001 1000 μL Pack of 1000
12150681 1000 μL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12029011 Supplier Mettler Toledo™ Supplier No. 17001133
BioClean™ tips provided in economical bulk bags for use with Rainin™ pipettes with LTS™ LiteTouch™.
  • Flawless performance: perfect production and advanced design
  • No deformations, cracks or flashes
  • 100% inert: purity and cleanliness are assured
  • All tips are 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors and additives
  • Packaged in a re-sealable zip-lock bag, for manual rack refilling
Specifications
Volume (Metric) 300 μL
Filtered Non-filtered
Sterility Non-sterile
Packaging 1000 Tips/Bag
For Use With (Equipment) Rainin LTS LiteTouch
Product Line Rainin BioClean