Learn More
Mettler Toledo™ Rainin™ BioClean™ Bulk Pipette Tips
Click to view available options
Volume (Metric):
300 μL
1000 μL
2000 μL
Unit Size:
Pack of 1000
Pack of 200
Description
- Flawless performance: perfect production and advanced design
- No deformations, cracks or flashes
- 100% inert: purity and cleanliness are assured
- All tips are 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors and additives
- Packaged in a re-sealable zip-lock bag, for manual rack refilling
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Packaging | 1000 Tips/Bag |
| For Use With (Equipment) | Rainin LTS LiteTouch |
| Product Line | Rainin BioClean |