Learn More
Mettler Toledo™ Rainin™ BioClean™ Bulk Pipette Tips
BioClean™ tips provided in economical bulk bags for use with Rainin™ pipettes with LTS™ LiteTouch™.
Brand:  Mettler Toledo™ 17001133
Additional Details : Weight : 2.36700kg
 View more versions of this product
Product Code. 12029011
  • POA / Pack of 1000
Estimated Shipment: 11-06-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
  • Flawless performance: perfect production and advanced design
  • No deformations, cracks or flashes
  • 100% inert: purity and cleanliness are assured
  • All tips are 100% free of DNA, DNAse, RNAse, ATP, Pyrogens, PCR inhibitors and additives
  • Packaged in a re-sealable zip-lock bag, for manual rack refilling
Specifications
Specifications
300 μL
Non-sterile
Rainin™ Pipette with LTS™ LiteTouch™
Non-Filtered
1000 tips/bag
Rainin BioClean