missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RalA Antibody (SR4327), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27797-100ul
This item is not returnable.
View return policy
Description
RalA Monoclonal antibody specifically detects RalA in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| RalA | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5898 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot | |
| SR4327 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| HINCONS, RAL | |
| A synthesized peptide derived from human RalA (Uniprot #: P11233) | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction