missing translation for 'onlineSavingsMsg'
Learn More

RALA Polyclonal Antibody, Invitrogen™

Product Code. 11521232
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11521232 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11521232 Supplier Invitrogen Supplier No. OSR00268W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 100 μL of distilled water.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RALA
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene RALA
Gene Accession No. P11233, P63321, P63322
Gene Alias Rala, Ral, Ral-a, MGC48949
Gene Symbols RALA
Host Species Rabbit
Immunogen A synthetic peptide from human Ras-related protein Ral-A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56044, 5898, 81757
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.