missing translation for 'onlineSavingsMsg'
Learn More

RalA/RalB Antibody (AT41E2), Biotin, Novus Biologicals™

Product Code. 18675730 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18675730 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18675730 Supplier Novus Biologicals Supplier No. NBP288840B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RalA/RalB Monoclonal antibody specifically detects RalA/RalB in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RalA/RalB
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone AT41E2
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA
Formulation PBS
Gene Alias MGC48949, RAL, Ras family small GTP binding protein RALA, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related)
Host Species Mouse
Immunogen Recombinant human RalA/RalB (1-203aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5898
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.