missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RalA/RalB Antibody (AT41E2), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88840MFV500
This item is not returnable.
View return policy
Description
RalA/RalB Monoclonal antibody specifically detects RalA/RalB in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| RalA/RalB | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| AT41E2 | |
| Flow Cytometry | |
| MGC48949, RAL, Ras family small GTP binding protein RALA, RAS-like protein A, ras-related protein Ral-A, v-ral simian leukemia viral oncogene homolog A (ras related) | |
| Recombinant human RalA/RalB (1-203aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 5898 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction