missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RalB Antibody (8A8A9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61826-0.025ml
This item is not returnable.
View return policy
Description
RalB Monoclonal specifically detects RalB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| RalB | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| GTP binding protein, RAS-like protein B, ras-related protein Ral-B, v-ral simian leukemia viral oncogene homolog B (ras related; GTP bindingprotein) | |
| Mouse | |
| 82.4 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA | |
| 8A8A9 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RALB | |
| Purified recombinant fragment of human RALB (AA: 89-206) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 5899 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction