missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RalB Antibody (OTI2C4), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73785DL594
This item is not returnable.
View return policy
Description
RalB Monoclonal antibody specifically detects RalB in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| RalB | |
| Monoclonal | |
| DyLight 594 | |
| GTP binding protein, RAS-like protein B, ras-related protein Ral-B, v-ral simian leukemia viral oncogene homolog B (ras related; GTP bindingprotein) | |
| Full length recombinant protein of human RalB (NP_002872) produced in HEK293T cells. | |
| 0.1 mL | |
| Signal Transduction | |
| 5899 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI2C4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction