missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALB Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI2C4, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505880
This item is not returnable.
View return policy
Description
This gene encodes a GTP-binding protein that belongs to the small GTPase superfamily and Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.Specifications
| RALB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11234, P36860, Q9JIW9 | |
| RALB | |
| Full length human recombit protein of human RALB produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| RALB | |
| RAS like proto-oncogene B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 116546, 483881, 5899, 64143 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction