missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RALB Polyclonal Antibody
GREENER_CHOICE

Product Code. 16383625
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16383625 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16383625 Supplier Invitrogen™ Supplier No. PA586762

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

This gene encodes a GTP-binding protein that belongs to the small GTPase superfamily and Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALB
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene RALB
Gene Accession No. P11234, P36860, Q9JIW9
Gene Alias 5730472O18Rik; dRalb; GTP binding protein; GTP binding protein); OTTHUMP00000204385; OTTHUMP00000207264; QtsA-18830; Ralb; Ral-b; RALB Ras like proto-oncogene B; RAS like proto-oncogene B; ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; unnamed protein product; v-ral simian leukemia viral oncogene homolog B; v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Symbols RALB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in Human RALB.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116546, 5899, 64143
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.