missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144683
This item is not returnable.
View return policy
Description
Immunogen sequence: PPDVVSDDEK DHGKKKGKFK KKEKRTEGYA AFQEDSSGDE AESPSKMKRS KGIHVFKKPS FSKKKEKDFK IKEKPKEEKH KEEKHKEEKH KEKKSKDLTA ADVVKQWKEK KKKKKPIQEP EVPQIDVPNL KP Antigen sequence identity: Mouse 95%; Rat 95%.
Can specifically activate hydrolysis of GTP bound to RAC1 and CDC42, but not RALA. Mediates ATP-dependent transport of S-(2,4-dinitrophenyl)-glutathione (DNP-SG) and doxorubicin (DOX) and is the major ATP-dependent transporter of glutathione conjugates of electrophiles (GS-E) and DOX in erythrocytes. Can catalyze transport of glutathione conjugates and xenobiotics, and may contribute to the multidrug resistance phenomenon. Serves as a scaffold protein that brings together proteins forming an endocytotic complex during interphase and also with CDC2 to switch off endocytosis, One of its substrates would be EPN1/Epsin.
Specifications
| RALBP1 | |
| Polyclonal | |
| Unconjugated | |
| RALBP1 | |
| 76 kDa Ral-interacting protein; Cytocentrin; dinitrophenyl S-glutathione ATPase; DNP-SG ATPase; ralA binding protein 1; ralA-binding protein 1; RALBP1; Ral-interacting protein 1; RIP; RIP1; RLIP1; RLIP76 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15311 | |
| RALBP1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 57-188. Recombinant protein control fragment (Product #RP-109972). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction