missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALGDS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100681
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RALGDS.
Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
Specifications
| RALGDS | |
| Polyclonal | |
| Unconjugated | |
| RALGDS | |
| Gnds; KIAA1308; mKIAA1308; Ral guanine nucleotide dissociation stimulator; ral guanine nucleotide exchange factor; RALGDS; RalGEF; RGDS; RGF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19730, 29622, 5900 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q03385, Q03386, Q12967 | |
| RALGDS | |
| A synthesized peptide derived from human RALGDS(Accession Q12967), corresponding to amino acid residues L160-E210. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction