missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALGPS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | RALGPS1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18657681
|
Novus Biologicals
NBP2-93902-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642711
|
Novus Biologicals
NBP2-93902-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RALGPS1 Polyclonal antibody specifically detects RALGPS1 in Human samples. It is validated for Western BlotSpecifications
| RALGPS1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9649 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| KIAA0351RalGEF 2, Ral GEF with PH domain and SH3 binding motif 1, Ral GEF with PH domain and SH3-binding motif 1, Ral guanine nucleotide exchange factor 2, Ral guanine nucleotide exchange factor RalGPS1A, RalA exchange factor RalGPS1, RALGEF2RALGPS1A, ras-specific guanine nucleotide-releasing factor RalGPS1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RALGPS1 (NP_055451.1). MYKRNGLMASVLVTSATPQGSSSSDSLEGQSCDYASKSYDAVVFDVLKVTPEEFASQITLMDIPVFKAIQPEELASCGWSKKEKHSLAPNVVAFTRRFNQVSFWVVREILTAQTLKIRAEILSHFVKIAKKLLELNNLHSLMSVVSALQSAPIFRLTKTWALLNRKDKTTFEKLDYLMSKEDNYKRTREYIRSLKMVPSIPYLGIYLLDLIYIDSAYPASGSIMENEQRSNQMNNILRIIADLQVSCSYDHLTTLPHVQKYLKSVRYIEELQKFVEDDNYKLSLRIEPGSSSPRLVSSKE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title