missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALGPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569173
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 76%.
RALGPS1 gene ontology annotations related to this gene include regulation of Ral protein signal transduction; small GTPase mediated signal transduction.
Specifications
| RALGPS1 | |
| Polyclonal | |
| Unconjugated | |
| RALGPS1 | |
| 5830418G11Rik; AI853783; AI854138; Kiaa0351; mKIAA0351; Ral GEF with PH domain and SH3 binding motif 1; ral GEF with PH domain and SH3-binding motif 1; Ral guanine nucleotide exchange factor 2; Ral guanine nucleotide exchange factor RalGPS1A; ralA exchange factor RalGPS1; RalGEF 2; RALGEF2; RALGPS1; Ralgps1a; ras-specific guanine nucleotide-releasing factor RalGPS1; si:dkey-191c17.1; zgc:165535 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9649 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5JS13 | |
| RALGPS1 | |
| synthetic peptide directed towards the middle region of human RALGPS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction