missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RALGPS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555823
This item is not returnable.
View return policy
Description
Immunogen sequence: VEDDNYKLSL KIEPGTSTPR SAASREDLVG PEVGASPQSG RKSVAAEGAL LPQTPPSPRN LIPHGHRKCH SLGYNFIHKM NTAEFKSATF PNAGP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 94%.
Specifications
| RALGPS2 | |
| Polyclonal | |
| Unconjugated | |
| RALGPS2 | |
| dJ595C2.1; Ral GEF with PH domain and SH3 binding motif 2; ral GEF with PH domain and SH3-binding motif 2; ralA exchange factor RalGPS2; Ral-A exchange factor RalGPS2; RALGPS2; Ras-specific guanine nucleotide-releasing factor RalGPS2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55103 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86X27 | |
| RALGPS2 | |
| Recombinant protein corresponding to Human RALGPS2. Recombinant protein control fragment (Product #RP-94064). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction