Learn More
Raltegravir, MedChemExpress
Product Code. 16480227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16480227
Brand: Medchem Express HY10353
MedChemExpress Raltegravir is a potent integrase (IN) inhibitor, used to treat HIV infection.
Raltegravir is a potent integrase (IN) inhibitor, used to treat HIV infection.
Specifications
Chemical Name or Material Raltegravir
CAS 518048-05-0
Color White
Purity Grade Notes Research
Molecular Formula C20H21FN6O5
Quantity 50 mg
Synonym MK-0518
Solubility Information DMSO : ≥ 100 mg/mL (225.01 mM)
SMILES O=C(C(O)=C(C(NCC1=CC=C(C=C1)F)=O)N=C2C(C)(C)NC(C3=NN=C(C)O3)=O)N2C
Molecular Weight (g/mol) 444.42
Formula Weight 444.42
Percent Purity 98.13%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only