missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALY Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97634JF646
This item is not returnable.
View return policy
Description
RALY Polyclonal specifically detects RALY in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| RALY | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| Autoantigen p542, Heterogeneous nuclear ribonucleoprotein C-like 2, hnRNP associated with lethal yellow protein homolog, hnRNP core protein C-like 2, HNRPCL2MGC117312, P542hnRNP-associated with lethal yellow), RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow), RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog(mouse)), RNA-binding protein (autoantigenic), RNA-binding protein Raly | |
| Produced in rabbits immunized with E. coli-derived Human RALY fragment. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 22913 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction