missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALY Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35195-100ul
This item is not returnable.
View return policy
Description
RALY Polyclonal antibody specifically detects RALY in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RALY | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| Autoantigen p542, Heterogeneous nuclear ribonucleoprotein C-like 2, hnRNP associated with lethal yellow protein homolog, hnRNP core protein C-like 2, HNRPCL2MGC117312, P542hnRNP-associated with lethal yellow), RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow), RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog(mouse)), RNA-binding protein (autoantigenic), RNA-binding protein Raly | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 96-306 of human RALY (NP_057951.1).,, Sequence:, DRPKGLKRAASAIYSGYIFDYDYYRDDFYDRLFDYRGRLSPVPVPRAVPVKRPRVTVPLVRRVKTNVPVKLFARSTAVTTSSAKIKLKSSELQAIKTELTQIKSNIDALLSRLEQIAAEQKANPDGKKKGDGGGAGGGGGGGGSGGGGSGGGGGGGSSRPPAPQENTTSEAGLPQGEARTRDDGDEEGLLTHSEEELEHSQDTDADDGALQ | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 22913 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction