missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ramiprilat (Ramipril Diacid), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Ramiprilat |
| CAS | 87269-97-4 |
| Analyte or Component Names | Ramiprilat |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H28 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | HOE 498 diacid, Ramipril diacid, Ramiprilat, Ramiprilate, Cyclopenta[b]pyrrole-2-carboxylic acid, 1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-1-oxopropyl]octahydro-, (2S,3aS,6aS)-, Cyclopenta[b]pyrrole-2-carboxylic acid, 1-[2-[(1-carboxy-3-phenylpropyl)amino]-1-oxopropyl]octahydro-, [2S-[1[R*(R*)],2alpha,3abeta,6abeta]]-, Ramipril Impurity E |
| InChI Formula | InChI=1S/C21H28N2O5/c1-13(22-16(20(25)26)11-10-14-6-3-2-4-7-14)19(24)23-17-9-5-8-15(17)12-18(23)21(27)28/h2-4,6-7,13,15-18,22H,5,8-12H2,1H3,(H,25,26)(H,27,28)/t13-,15-,16-,17-,18-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?