missing translation for 'onlineSavingsMsg'
Learn More

Ramiprilat, TRC

Product Code. 18040420
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040420 2.5 mg 2.5mg
18073510 5 mg 5mg
18070610 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18040420 Supplier Toronto Research Chemicals Supplier No. TRCR1110102.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87269-97-4
Molecular Formula C21 H28 N2 O5
Molecular Weight (g/mol) 388.46
Synonym Cyclopenta[b]pyrrole-2-carboxylic acid, 1-[2-[(1-carboxy-3-phenylpropyl)amino]-1-oxopropyl]octahydro-, [2S-[1[R*(R*)],2α,3aβ,6aβ]]-, HOE 498 diacid, Ramipril diacid, Ramiprilat, Ramiprilate, Cyclopenta[b]pyrrole-2-carboxylic acid, 1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]-1-oxopropyl]octahydro-, (2S,3aS,6aS)-
IUPAC Name (2S,3aS,6aS)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]-3,3a,4,5,6,6a-hexahydro-2H-cyclopenta[b]pyrrole-2-carboxylic acid
SMILES C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2[C@H]3CCC[C@H]3C[C@H]2C(=O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H28N2O5/c1-13(22-16(20(25)26)11-10-14-6-3-2-4-7-14)19(24)23-17-9-5-8-15(17)12-18(23)21(27)28/h2-4,6-7,13,15-18,22H,5,8-12H2,1H3,(H,25,26)(H,27,28)/t13-,15-,16-,17-,18-/m0/s1
Formula Weight 388.1998
Percent Purity >95%
Chemical Name or Material Ramiprilat
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.