missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP2 Antibody (773816) [DyLight 488], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6500K
This item is not returnable.
View return policy
Description
RAMP2 Monoclonal antibody specifically detects RAMP2 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| RAMP2 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10266 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 773816 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
| E. coli-derived recombinant mouse RAMP2, Ser45-Val159, Accession # Q9WUP0 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction