missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAMP2 Antibody (OTI4E5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73796AF532
This item is not returnable.
View return policy
Description
RAMP2 Monoclonal specifically detects RAMP2 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| RAMP2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RAMP2 | |
| Full length recombinant protein of human RAMP2 (NP_005845) produced in HEK293T cells. | |
| 0.1 mL | |
| 10266.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4E5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction