missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP1 Monoclonal Antibody (OTI8E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811554
This item is not returnable.
View return policy
Description
Ran/TC4-binding protein, RanBP1, interacts specifically with GTP-charged RAN. RANBP1 encodes a 23-kD protein that binds to RAN complexed with GTP but not GDP. RANBP1 does not activate GTPase activity of RAN but does markedly increase GTP hydrolysis by the RanGTPase-activating protein (RanGAP1). The RANBP1 cDNA encodes a 201-amino acid protein that is 92% similar to its mouse homolog. In both mammalian cells and in yeast, RANBP1 acts as a negative regulator of RCC1 by inhibiting RCC1-stimulated guanine nucleotide release from RAN.Specifications
| RANBP1 | |
| Monoclonal | |
| Unconjugated | |
| RANBP1 | |
| HTF9A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5902 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8E1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43487 | |
| RANBP1 | |
| Full length human recombit protein of human RANBP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction