missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP1 Monoclonal Antibody (OTI8E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811554S
This item is not returnable.
View return policy
Description
RANBP1 Monoclonal antibody specifically detects RANBP1 in Human samples. It is validated for Western BlotSpecifications
| RANBP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41439 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2352 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI8E1 | |
| Unconjugated | |
| FOLR3 | |
| HpaII tiny fragments locus 9A; hpaII tiny fragments locus 9a protein; Htf9a; Htf9-a; Htf9-a/RanBP1; RAN binding protein 1; ran-binding protein 1; RANBP1; RANG; Ran-specific GTPase-activating protein; testis secretory sperm-binding protein Li 221n | |
| Full length human recombinant protein of human FOLR3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction