missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP16 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18480V
This item is not returnable.
View return policy
Description
RanBP16 Polyclonal antibody specifically detects RanBP16 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RanBP16 | |
| Polyclonal | |
| Western Blot | |
| EXP7, exportin 7, KIAA0745exportin-7, RAN binding protein 16, Ran-binding protein 16, RANBP16Exp7 | |
| Recognizes region between residues Between between 1 and 50.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23039 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction