missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RanBP16 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 630.00
Specifications
| Antigen | RanBP16 |
|---|---|
| Concentration | 0.2 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17169103
|
Invitrogen™
PA5111187 |
100 μL |
€ 630.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17179183
![]() |
Invitrogen™
PA5111268 |
100 μL |
€ 630.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: SQPPEKQQAM HLCFENLMEG IERNLLTK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
The transport of protein and large RNAs through the nuclear pore complexes is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN plays a key role in NLS-dependent protein import. RAN-binding protein-16 is a member of the importin-beta superfamily of nuclear transport receptors.Specifications
| RanBP16 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| Xpo7 | |
| 4930506C02Rik; BB164534; EXP7; exportin 7; exportin-7; Kiaa0745; mKIAA0745; RAN binding protein 16; ran-binding protein 16; Ranbp16; Xpo7 | |
| Xpo7 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UIA9 | |
| 23039 | |
| Recombinant Protein corresonding to Human RanBP16. Recombinant protein control fragment (Product #RP-100395). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title