missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RanBP3 Antibody (12E11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42672AF405
This item is not returnable.
View return policy
Description
RanBP3 Monoclonal specifically detects RanBP3 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| RanBP3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RANBP3 | |
| Recombinant human RANBP3 (235-445aa) purified from E. coli | |
| 100 μL | |
| Signal Transduction | |
| 8498 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1200000000000 | |
| Western Blot, ELISA | |
| DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction