missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RANBP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584120
This item is not returnable.
View return policy
Description
Immunogen sequence: LSSGFEEVYP NLPADVQRDV KIELILAVKL ETHA Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 71%.
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.
Specifications
| RANBP6 | |
| Polyclonal | |
| Unconjugated | |
| RANBP6 | |
| C630001B19; RAN binding protein 6; Ran BP6; ran-binding protein 6; Ranbp6; Ran-BP6; Ran-GTP binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26953 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60518 | |
| RANBP6 | |
| Recombinant protein corresponding to Human RANBP6. Recombinant protein control fragment (Product #RP-106408). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction