missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP9 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98142AF647
This item is not returnable.
View return policy
Description
RANBP9 Polyclonal antibody specifically detects RANBP9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RANBP9 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| BPM90, BPM-L, RAN binding protein 9, Ran Binding Protein in the Microtubule organizing center, ran binding protein, centrosomal, ran-binding protein 9, Ran-binding protein M, RanBP7, RanBP9, RanBPM, RANBPMnovel centrosomal protein RanBPM | |
| Produced in rabbits immunized with E. coli-derived Human RANBP9 fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 10048 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction