missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANBP9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93772-0.1ml
This item is not returnable.
View return policy
Description
RANBP9 Polyclonal antibody specifically detects RANBP9 in Human samples. It is validated for Western Blot
Specifications
| RANBP9 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| BPM90, BPM-L, RAN binding protein 9, Ran Binding Protein in the Microtubule organizing center, ran binding protein, centrosomal, ran-binding protein 9, Ran-binding protein M, RanBP7, RanBP9, RanBPM, RANBPMnovel centrosomal protein RanBPM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 480-605 of human RanBP9 (NP_005484.2). PFSSPSMSPSHGMNIHNLASGKGSTAHFSGFESCSNGVISNKAHQSYCHSNKHQSSNLNVPELNSINMSRSQQVNNFTSNDVDMETDHYSNGVGETSSNGFLNGSSKHDHEMEDCDTEMEVDSSQL | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 10048 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction