Learn More
R&D Systems™ Polystyrene Microplates
Click to view available options
Color:
Clear
Unit Size:
Pack of 25
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Clear |
| Product Type | ELISA Microplate |
| Material | Polystyrene |
| Volume (Metric) Well | 360 μL |
| No. of Wells | 96 |
| Well Shape | Flat Bottom (F-well) |
| For Use With (Application) | ELISA |