Learn More
- Home
- Products
- Personal Protective Equipment
- Foot Protection
- General Purpose Work Boots and Shoes
- Emma Safety Footwear Ranger Nubuck Leather Outdoor D-Fit Shoes
Emma Safety Footwear Ranger Nubuck Leather Outdoor D-Fit Shoes
Product Code. 13276484
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Each
32 product options available for selection
| Product Code. | Size | Unit Size |
|---|---|---|
| 13276484 | 44 | 1 pair |
| 13286484 | 45 | 1 pair |
| 10883231 | 45 | Each |
| 10387604 | 45 | 1 pair |
| 10565190 | 36 | Each |
| 10422094 | 36 | 1 pair |
| 13296474 | 36 | 1 pair |
| 10684605 | 48 | 1 pair |
| 13284404 | 48 | 1 pair |
| 11337080 | 48 | Each |
| 13216484 | 38 | 1 pair |
| 10103884 | 38 | 1 pair |
| 13236484 | 40 | 1 pair |
| 10348949 | 40 | Each |
| 10255254 | 40 | 1 pair |
| 13246484 | 41 | 1 pair |
| 10634415 | 41 | 1 pair |
| 10767515 | 42 | 1 pair |
| 13256484 | 42 | 1 pair |
| 10193974 | 43 | 1 pair |
| 13266484 | 43 | 1 pair |
| 10082474 | 44 | 1 pair |
| 13264404 | 46 | 1 pair |
| 10614255 | 46 | 1 pair |
| 10799625 | 47 | 1 pair |
| 13274404 | 47 | 1 pair |
| 13206484 | 37 | 1 pair |
| 10317374 | 37 | 1 pair |
| 13226484 | 39 | 1 pair |
| 10245254 | 39 | 1 pair |
| 10412094 | 35 | 1 pair |
| 13286474 | 35 | 1 pair |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
This item is not returnable.
View return policy
Product Code.
13276484
Supplier
Emma Safety Footwear
Supplier No.
MM76054644
This item is not returnable.
View return policy
Description
- Nubuck/Cordura™ uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifications
Specifications
| Liner Material | Hydro-Tec sanitized silver lining |
| Size | 44 |
| Sole Features | Non-woven antistatic insole |
| Sole Material | GripForce™ Easy Twist outsole |
| Toe Style | Steel toe |
| Product Type | Nubuck Leather Outdoor Shoes |