Learn More
Ranitidine-d6 HCl (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ranitidine-d6 Hydrochloride |
| CAS | 1185238-09-8 |
| Molecular Formula | C13 2H6 H16 N4 O3 S . Cl H |
| Quantity | 0.01 g |
| Synonym | Ranitidine-D6 Hydrochloride |
| InChI Formula | InChI=1 S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15 H,6-8,10H2,1-3H3;1 H/i2D3,3D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(Cc1oc(CSCCNC(=C[N+](=O)[O-])NC)cc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-N'-[2-[[5-[[bis(trideuteriomethyl)amino]methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride |
| Molecular Weight (g/mol) | 356.9 |
| Formula Weight | 356.1556 g/mol |
| Show More |