Learn More
Invitrogen™ RANK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110945
Description
Immunogen sequence: CTTTSDSVCL PCGPDEYLDS WNEEDKCLLH KVCDTGKALV AVVAGNSTTP RRCACTAGYH WSQDCECCRR NTECAPGLGA QHPLQLNKDT VCKPCLAGYF SDAFSSTDKC RPWTNCTFLG KRVEHHGTEK SDAVCSSSLP ARKPPNEPHV Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 79%.
RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
Specifications
| RANK | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF11A | |
| CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8792 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6Q6 | |
| TNFRSF11A | |
| Recombinant Protein corresonding to Human RANK. Recombinant protein control fragment (Product #RP-90287). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.