missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody (107), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90324C
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human samples. It is validated for ELISA.
Specifications
| RANK/TNFRSF11A | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 107 | |
| ELISA | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human RANK/TNFRSF11A (NP_001257879.1; Met1-Pro212). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8792 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction