missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody (64C1385.1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-24702F
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| RANK/TNFRSF11A | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF11A | |
| A recombinant protein containing the extracellular domain (amino acids 33-208) of human RANK was used as immunogen. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8792 | |
| Store at 4C in the dark. | |
| IgG1 λ |
| Flow Cytometry | |
| 64C1385.1 | |
| Flow Cytometry | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction