missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody (9A725), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56508B
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Monoclonal specifically detects RANK/TNFRSF11A in Human, Mouse, Rat, Avian samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RANK/TNFRSF11A | |
| Monoclonal | |
| Biotin | |
| Q9Y6Q6 | |
| TNFRSF11A | |
| A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8792 | |
| Human, Mouse, Rat, Avian | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 9A725 | |
| Western Blot, Immunohistochemistry-Paraffin | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| In RAW cells, a 97 kDa is observed. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction