missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RANK/TNFRSF11A Antibody (9A725), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56508MFV500
This item is not returnable.
View return policy
Description
RANK/TNFRSF11A Monoclonal antibody specifically detects RANK/TNFRSF11A in Human, Mouse, Rat, Avian samples. It is validated for Flow Cytometry, Flow (Cell Surface)
Specifications
| RANK/TNFRSF11A | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Avian | |
| Purified |
| Flow Cytometry | |
| 9A725 | |
| Flow Cytometry, Flow (Cell Surface) | |
| CD265, CD265 antigen, FEO, loss of heterozygosity, 18, chromosomal region 1, ODFROSTS, OFE, OPTB7, Osteoclast differentiation factor receptor, PDB2, RANKLOH18CR1, Receptor activator of NF-KB, receptor activator of nuclear factor-kappa B, TRANCER, tumor necrosis factor receptor superfamily member 11A, tumor necrosis factor receptor superfamily, member 11a, activator of NFKB, tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
| A fusion protein containing amino acid residues 326-616 of human RANK was used as immunogen. This antibody recognizes an epitope located between amino acid residues 330-427 of human RANK (NP_003830). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8792 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction