missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP Polyclonal Antibody
RAP Polyclonal Antibody
Brand: Invitrogen PA547626
This item is not returnable.
View return policy
Description
RAP Polyclonal Antibody for Western Blot
Specifications
| RAP | |
| Polyclonal | |
| Unconjugated | |
| Lrpap1 | |
| 39 kDa receptor-associated protein; A2MRAP; A2RAP; AA617339; AI790446; Alpha-2-macroglobulin receptor-associated protein; Alpha-2-macroglobulin receptor-associated protein precursor (Alpha-2-MRAP) (Low density lipoprotein receptor-related protein-associated protein 1) (RAP) (Heparin binding protein-44) (HBP-44); alpha-2-MRAP; AU042172; C77774; gp330-binding 45 kDa protein; HBP44; HBP-44; heparin-binding protein 44; LDL receptor related protein associated protein 1; low density lipoprotein receptor related protein, associated protein 1; low density lipoprotein receptor-related protein associated protein 1; low density lipoprotein receptor-related protein-associated protein 1; low density lipoprotein-related protein-associated protein 1 (alpha-2-macroglobulin receptor-associated protein 1); Lrpap1; MRAP; MYP23; RAP | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16976 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P55302 | |
| Lrpap1 | |
| E. coli-derived recombinant mouse LRPAP Gln29-Leu360. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction